Hamilton Removable Syringe Needle, Small Hub, Conical Tip, 26s G; 6/Pk
Representative image only.
Hamilton Mfr # 7784-07 – Item # WW-25703-80
Ideal for chromatography
  • Constructed of durable 304 stainless steel
  • Autoclavable
  • Needle point style 5 features a conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring
  • Small hub removable (RN) needles can be used with removable needle syringes that are 100 μL and smaller
  • 1 year warranty
PRODUCT OPTIONS
₹17,400.00INR / Pkg of 6
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeSmall hub removable needle
  • Needle Tip Number5
  • Needle Tip StyleConical
  • Needle Gauge26s
  • Needle Length (in)2
  • Material304 stainless steel
  • DescriptionRemovable Syringe Needle, Small Hub, Conical Tip, 26s G; 6/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Constructed of durable 304 stainless steel
  • Autoclavable
  • Needle point style 5 features a conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring
  • Small hub removable (RN) needles can be used with removable needle syringes that are 100 μL and smaller
  • 1 year warranty
More About this Item
Hamilton® removable syringe needles offer the benefit of a replaceable needle for the occasional bend or clog and the minimal dead volume of a cemented needle. The unique removable needle design installs the needle precisely at the zero line.