Hamilton Gastight® SampleLock™ Syringe, Large Hub Removable Needle, Beveled Tip, 500 µL, 22 G
Representative image only.
Hamilton Mfr # 81256 – Item # WW-07940-55
Gastight sample locking up to 500 psi
  • Gastight syringes feature an on/off valve needle
  • Needle point style 2 features a sharp, beveled, curved, non-coring needle point recommended for piercing septa
  • Syringe can handle up to 700 psi max pressure
  • 1 year warranty
PRODUCT OPTIONS
₹19,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeSampleLock™ Needle
  • Syringe Capacity (µL)500
  • Needle IncludedYes
  • Needle Tip StyleSharp, beveled, curved, non-coring
  • Needle Tip Number2
  • Needle Gauge22
  • Needle Length (in)2
  • MaterialSST, Borosilicate glass, PTFE
  • Outside Diameter (OD) (mm)7.75
  • DescriptionSampleLock™ Syringe, Large Hub Removable Needle, Beveled Tip, 500 µL, 22 G
  • Warranty1 year
1 Year Warranty
Key Features
  • Gastight syringes feature an on/off valve needle
  • Needle point style 2 features a sharp, beveled, curved, non-coring needle point recommended for piercing septa
  • Syringe can handle up to 700 psi max pressure
  • 1 year warranty
More About this Item
The Hamilton® Gastight® SampleLock™ syringes allow you to collect, store, transport and analyze liquid or gaseous samples without the danger of evaporative loss or environmental contamination. An easy-to-use twist valve and a positive rear stop on SampleLock prevents loss of sample and plunger blowout.
SampleLock™ Syringes incorporate an on/off valve with removable needle. Use these syringes for headspace, environmental sample collection and storage, prepressurization of gaseous samples for GC analysis, and sample spiking. Not autoclavable.Use the sharp, curved, noncoring needle for septum penetration applications such as gas chromatography.