Hamilton Gastight® Syringe with Cemented Needle, Blunt Tip, 100 µL, 22s G
Representative image only.
Hamilton Mfr # 81075 – Item # WW-07940-37
Remain gas- and liquid-tight!
  • Syringes handle corrosive gases and liquids, radioactive materials, and sterile solutions—ideal for gas and liquid chromatography
  • Durable epoxy-cemented stainless steel needle
  • Dead volume is limited to the internal volume of the needle
  • Needle point style 3 features a blunt needle point for use with HPLC injection valves and for sample pipetting.
  • 1 year warranty
PRODUCT OPTIONS
₹7,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeCemented Needle
  • Syringe Capacity (µL)100
  • Needle Tip Number3
  • Needle Tip Style90° Blunt
  • Needle IncludedYes
  • Needle Gauge22s
  • Needle Length (in)2
  • MaterialSST, Borosilicate glass, PTFE
  • SterileNo
  • Outside Diameter (OD) (mm)7.75
  • DescriptionSyringe with Cemented Needle, Blunt Tip, 100 µL, 22s G
  • Warranty1 year
1 Year Warranty
Key Features
  • Syringes handle corrosive gases and liquids, radioactive materials, and sterile solutions—ideal for gas and liquid chromatography
  • Durable epoxy-cemented stainless steel needle
  • Dead volume is limited to the internal volume of the needle
  • Needle point style 3 features a blunt needle point for use with HPLC injection valves and for sample pipetting.
  • 1 year warranty
More About this Item
Hamilton® Gastight® syringes with cemented needle are constructed with a chemically inert glass barrel. Gastight syringes are ideal for dispensing both liquids and gases. They have a precision-machined PTFE plunger tip which creates a leak-free seal.
Choose from cemented needle which includes an epoxy-cemented stainless steel needle or luer-tip cemented needle which features an epoxy-cemented needle on a luer tip for added stability.