Hach TNT877 TNTplus® Spectrophotometer Chemistry, Boron, 0.05 to 2.50 mg/L Boron; 25/PK
Representative image only.
Hach Mfr # TNT877 – Item # WW-99515-53
Perform common methods with these pre-made reagent tests
  • Measures: Boron, 0.05 to 2.50 mg/L Boron
PRODUCT OPTIONS
₹44,500.00INR / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • MaterialBoron, 0.05 to 2.50 mg/L Boron
  • Compatible WithHach DR 6000, 3900, 1900
  • DescriptionSpectrophotometer Chemistry, Boron, 0.05 to 2.50 mg/L Boron; 25/PK
Key Features
  • Measures: Boron, 0.05 to 2.50 mg/L Boron
More About this Item
Dozens of options to cover most applications!