Hach 2822800 Arsenic EZ High Range Test Kit
Representative image only.
Hach Mfr # 2822800 – Item # WW-05546-12
Reduce risk by using as a pre-test validation tool
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
SPECS & DESCRIPTION
Specifications & Description
  • TestArsenic
  • Measurement Points0, 10, 25, 50, 100, 250, 500; 0, 35, 75, 175, 1500, 4000 ppb
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionArsenic Ez Dual Range Test Strips, 100
ISO9001 Certified Supplier
Includes
three reagents, test strips, reaction vessel and cap, cotton balls, sample cell, instructions and carrying case.
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Dual range Arsenic test kit from 0 to 500 ppb and 0 to 4000 ppb, and able to preform 100 tests.