H-B Instrument B60380-0500 Easy-Read Liquid-In-Glass Tank Thermometer, 0 to 230°F, 76 mm Immersion, Environmentally Friendly

Representative image only.
H-B Instrument Mfr # 603800500 – Item # WW-93902-05
Supplied with ASTM "Like" approved, non-mercury thermometers
  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Supplied with ASTM "Like" approved, non-mercury thermometers
  • Thermometers can be remounted can be replaced if breakage occurs
  • 1 year warranty

PRODUCT OPTIONS

Call For Price

₹27,000.00 - ₹33,700.00INR / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Min Temperature (° F)0
  • Length (\")16
  • Min Temperature (° C)-18
  • Max Temperature (° F)230
  • Max Temperature (° C)110
  • Accuracy±2°F
  • Division (° F)2
  • Thermometer FillBlack Spirit
  • Dial / Case MaterialWood with Brass Cup
  • Overall Length (in)16
  • Overall Length (mm)405
  • Immersion Depth (mm)76
  • Stem MaterialYellow-Back Glass
  • DescriptionEasy-Read Liquid-In-Glass Tank Thermometer; 0 to 230F, 76mm Immersion, Environmentally Friendly
  • Warranty1 year
1 Year Warranty

Key Features

  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Supplied with ASTM "Like" approved, non-mercury thermometers
  • Thermometers can be remounted can be replaced if breakage occurs
  • 1 year warranty

More About this Item

All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant and have a statement of accuracy.

Wood/Brass cup case is designed with a non-sparking brass cup which holds which holds 20ml of liquid, which meets with both ASTM and API requirements. The thermometer is held in place with spring clips. The hardwood is lacquered to prevent absorption from the sample

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat