Guardian Equipment G1898LH-L AutoFlow Wall-Mount Eyewash, Stainless Steel
Representative image only.
Guardian Equipment Mfr # G1898LH-L – Item # WW-06767-13
Simply pull down the sprayhead to activate flow
  • Inline strainer protects sprayheads from debris in the water line
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Eyewash Flow Rate (GPM)3.6
  • Eyewash Fixture MaterialStainless steel
  • MountWall Mount
  • Inlet Connection1/2\" NPT(F)
  • ComplianceANSI Z385.1-2009
  • DescriptionAutoFlow Wall-Mount Eyewash, Stainless Steel
  • Warranty2 year
Two year warranty
Key Features
  • Inline strainer protects sprayheads from debris in the water line
  • 2 year warranty
More About this Item
Install this wall-mount eyewash above your sink or install above a floor drain to prevent water accumulation. Features two GS-Plus™ sprayheads with a flip-top dust cover, internal flow control, and filter. Eyewash actuates upon pulling the assembly down and remains in operation until it is pushed back into the storage position. Water supply inlet is 1/2" NPT(F).
Compliance: ANSI Z385.1-2009