Graphic Controls 8" Circular Chart Recorder Paper, 5 to 40°C / -20 to 120 %RH, 7 Days, 60/Pack
Representative image only.
Graphic Controls Mfr # 00553669 – Item # WW-80013-17
Standard chart recorder paper compatible with common brands
  • Circular or strip sheets
  • Fits Dickson recorder models CO2X, ESX, PR8, PRXA, PRPR8, TH8, and THDX
  • 1 year warranty
PRODUCT OPTIONS
₹7,200.00INR / Pkg of 60
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeCircular
  • Chart Speed7 day
  • Chart Diameter (in)8
  • Max Temperature (° C)40
  • Min Temperature (° C)5
  • Max Humidity (RH%)120
  • Min Humidity (RH%)-20
  • Description8\" Circular Chart Recorder Paper, 5 to 40°C / -20 to 120 %RH, 7 Days, 60/Pack
  • Warranty1 year
1 Year Warranty
Key Features
  • Circular or strip sheets
  • Fits Dickson recorder models CO2X, ESX, PR8, PRXA, PRPR8, TH8, and THDX
  • 1 year warranty
More About this Item
Choose the size and type that meets your recording needs.