Graphic Controls 8" Chart Paper, 0 to 100, 24 Hour; 60/Pk
Representative image only.
Graphic Controls Mfr # 00166132 – Item # WW-80013-02
Monitor with precision accuracy for quality control
  • Only authentic Dickson charts and pens guarantee accuracy
  • Dickson charts are a great way to archive your monitoring history
  • Flexible options to choose time, temperature, and size
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeCircular
  • Chart Speed24 hour
  • Chart Diameter (in)8
  • Min Temperature (° F)0
  • Max Temperature (° C)100
  • Min Temperature (° C)-18
  • Description8\" Chart Paper, 0 to 100, 24 Hour; 60/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Only authentic Dickson charts and pens guarantee accuracy
  • Dickson charts are a great way to archive your monitoring history
  • Flexible options to choose time, temperature, and size
  • 1 year warranty
More About this Item
Choose the paper to meet your application needs.