Global Water WQ101 Temperature Sensor, Stainless Steel, 4-20 Ma Output
Representative image only.
Global Water Mfr # 400760-025 – Item # WW-99243-32
Rugged, reliable sensors handle your open water environmental monitoring applications
  • Encapsulated electronics and stainless steel housing are fully submersible
  • 4 to 20 mA output is compatible with most displays, dataloggers, and logic controllers
  • Sensors are available to measure most common water quality parameters including pH, ORP, conductivity, DO, turbidity, and temperature
  • 1 year warranty
₹189,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Control TypeOn/Off
  • Length (cm)11.43
  • Diameter (cm)1.91
  • Length (in)4 1/2
  • Diameter (in)3/4
  • DescriptionTemperature Sensor, Stainless Steel, 4-20 Ma Output
  • Warranty1 year
1 Year Warranty
Key Features
  • Encapsulated electronics and stainless steel housing are fully submersible
  • 4 to 20 mA output is compatible with most displays, dataloggers, and logic controllers
  • Sensors are available to measure most common water quality parameters including pH, ORP, conductivity, DO, turbidity, and temperature
  • 1 year warranty
More About this Item
Global Water temperature sensors are rugged and reliable measurement devices ideal for submersible measurement applications such as stream and lake monitoring, or aquaculture studies. Fully encapsulated electronics allow for full submersion without damage to sensor internals. Stainless steel housing resists corrosion and physical damage. Marine-grade cables offer additional environmental resistance.
All units feature a 4 to 20 mA output which is accepted by most local displays, dataloggers, remote telemetry units, and logic controllers. Optional sensor display offers local readout on a large 4-digit LCD. Optional datalogger monitors up to 9 different sensors, records over 40,000 readings, and exports data via USB, DB9, or RS-232 ports. Order display and datalogger separately.
Full specifications can be viewed by downloading the instruction manual