Global Specialties 105-9000 Multi-Tester RF Generator 4-In-1
Representative image only.
Global Specialties Mfr # 105-9000 – Item # WW-20018-71
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMulti-Tester RF Generator 4-In-1
More About this Item
FEATURES
  • Four products in one package with none of the functions of each individual product compromised
  • 100KHz to 110MHz RF generator with AM & FM capabilities
  • 100MHz high sensitivity internal/external frequency counter with 5 digit display
  • 1Hz to 1MHz function generator
  • Adjustable voltage and current limit
  • Digital readout for voltage and current 32VDC @ 10A
APPLICATIONS INCLUDE:
  • R&D labs
  • Test bench testing at industrial and electronic manufacturers, service & repair facilities
  • Special RF testing and repair work
  • Education institutions as they set up new compu