GF Signet 3-2821-1 2821 Conductivity Electrode; SS, K=1.0, 3/4"NPT(M)
Representative image only.
GF Signet Mfr # 3-2821-1 – Item # WW-19905-51
Coupled with +GF+ Signet measuring circuitry, a three decade measurement range reading is achieved without the need for troublesome electrode platinization
  • Flow-through design eliminates bubble entrapment and sediment build-up
  • Removable/reversible sensor fitting design
  • Built-in strain relief
  • Controlled surface finish ensures accuracy and repeatability
  • In-line or submersible mounting
  • 1 year warranty
PRODUCT OPTIONS
₹81,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Cell ConstantK = 1.0
  • Cell TypeContact
  • Electrode MaterialPlatinum
  • Body Material316 SS
  • Fitting¾\" NPT(M)
  • Mounting TypeThreaded
  • Min temperature (° C)0
  • Max temperature (° C)55
  • Min temperature (° F)32
  • Max temperature (° F)130
  • Cable Length (m)4.6
  • Cable Length (ft)15
  • Description2821 Conductivity Electrode; SS, K=1.0, 3/4\"NPT(M)
  • Warranty1 year
1 Year Warranty
Key Features
  • Flow-through design eliminates bubble entrapment and sediment build-up
  • Removable/reversible sensor fitting design
  • Built-in strain relief
  • Controlled surface finish ensures accuracy and repeatability
  • In-line or submersible mounting
  • 1 year warranty
More About this Item
+GF+ Signet Conductivity/Resistivity Electrodes are designed to provide versatile installation and accurate sensing across a very broad dynamic range. Platinum RTD (PT-1000) located within the electrode allows optimal temperature sensing.