GF Signet 3-2726-HF-10 pH Electrode, HF Resistant, 3 kOhm Balco Thermistor, 3/4" NPT Threads
Representative image only.
GF Signet Mfr # 3-2726-HF-10 – Item # WW-56561-06
Unique DryLoc design provides maximum installation flexibility
  • Chemically resistant Ryton® body for use in dirty or corrosive environments
  • Easy retrofitting! Mounts to existing off-the-shelf and +GF+ Signet installation fittings
  • HF-resistant bulb pH electrode with 3/4" NPT (M) thread and 3 kΩ Balco ATC
  • 6 month warranty
PRODUCT OPTIONS
₹128,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsHF resistant
  • Electrode Body MaterialRyton® PPS
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)10
  • Max Temperature (° C)80
  • Temperature CompensationYes
  • Max Pressure (PSI)100
  • Thread Size NPT(M) (in)3/4
  • ATC Element3 kΩ Balco
  • ConnectorDryLoc (ISO 7/1 R3/4)
  • DescriptionpH Electrode, bulb, HF resistant, 3KOhm Balco thermistor, 3/4\" NPT threads
  • Warranty6 month warranty
6 Month Warranty
Key Features
  • Chemically resistant Ryton® body for use in dirty or corrosive environments
  • Easy retrofitting! Mounts to existing off-the-shelf and +GF+ Signet installation fittings
  • HF-resistant bulb pH electrode with 3/4" NPT (M) thread and 3 kΩ Balco ATC
  • 6 month warranty
More About this Item
Electrodes use a unique foul-proof DryLoc connector with gold plated contacts for extended lifetime. Choice of flat pH, bulb pH, and flat ORP sensing elements provides for a broad range of uses. HF resistant versions are for use in applications where traces of hydrofluoric acid will attack standard glass. LC versions are designed for use in low conductivity fluids and ultrapure water applications.
DryLoc connectors or preamplifiers are required for initial installation. Order connectors and preamplifiers separately.