GF Signet 155.522.554 PVDF Needle Valve, 1/4" NPT(F), with PTFE
Representative image only.
GF Signet Mfr # 155.522.554 – Item # WW-01369-20
₹33,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.25
  • Outlet Port1/4" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/4" NPT(F)
  • Max Pressure (PSI)200
  • Max Pressure (bar)200
  • Max Temperature (° F)302
  • Max Temperature (° C)150
  • Wetted MaterialsPVDF
  • Working Pressure (PSI)13.8
  • DescriptionPVDF Needle Valve, 1/4" NPT(F), with PTFE
More About this Item
PVDF needle valves are pressure rated up to 200 psi (13.8 bar) with a max temperature rating of 302 deg F (150 deg C). Seats are made of PTFE and O-rings made of Viton®. Valves feature standard pipe thread connections and are autoclavable.