George Ulanet 290-1 HeetGrid Immersion Heater with Dial Controller, 1000 watts, 7" dia, 230 VAC
Representative image only.
George Ulanet Mfr # 290-1 – Item # WW-03047-12
  • 6 Month Product / 12 Month Parts and Labor Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Immersion Depth (in)5
  • Heated Area5 in L
  • Max Temperature (° F)212
  • Max Temperature (° C)100
  • Min TemperatureAmbient;Ambient
  • Plug TypeNo plug
  • Power (VAC)230
  • Power (watts)1000
  • DescriptionHeetGrid Immersion Heater with Dial Controller, 1000 watts, 7\" dia, 230 VAC
  • Warranty6 Month Product / 12 Month Parts and Labor
Key Features
  • 6 Month Product / 12 Month Parts and Labor Warranty
More About this Item
Heaters with Dial Temperature Controller have a sensitivity of ±1°>F (±0.6°>C) in an agitated bath. Features a coiled heating element to concentrate heat at the bottom of the vessel. Heaters include a 6-ft, three-wire cord with plug on 115 VAC models; no plug on 230 VAC models.