General Tools Magnetic Tool with 2-lb. Pick-up Capacity
Representative image only.
General Tools Mfr # 386 – Item # WW-09914-30
Increase retrieval potential with a magnetic reach
  • 1 year warranty
PRODUCT OPTIONS
₹1,400.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Adjustable Handle Length5-1/2\"
  • DescriptionMagnetic Tool with 2-lb. Pick-up Capacity
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These magnetic tools feature high power neodymium magnets that are six times more powerful than a standard cobalt magnet. Compact size allows these tools to fit in your pocket. For greater reach, models 09914-34, -36, and –38 feature extendable shafts for those out-of-reach objects.