General Tools & Instruments PS10 Pressure Transmitter for 68602-20, 0 to 145 psi
Representative image only.
General Tools & Instruments Mfr # PS10 – Item # WW-68602-26
Measure a variety of pressure ranges with a single meter
  • Settable parameters of date, time, auto power off, beep sound, sampling time, decimal point or comma decimal division
  • Data logging sampling time settable from 1 second to 9 hours
  • Minimum and maximum recallable readings
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPressure Transmitter for 68602-20, 0 to 145 psi
  • Warranty1 year
1 Year Warranty
Key Features
  • Settable parameters of date, time, auto power off, beep sound, sampling time, decimal point or comma decimal division
  • Data logging sampling time settable from 1 second to 9 hours
  • Minimum and maximum recallable readings
  • 1 year warranty
More About this Item
This versatile meter, along with the interchangeable transducers, provides a great solution for pressure measurement requirements in HVAC/R, industrial, and process applications. The transducers connect to the meter via a 4-pin M12 electronic connection with a 3-ft (0.9-m) long cable. The 2 GB SD card stores data in Excel® format for logging and downloading onto your computer for record keeping.
Note: Interchangeable pressure tranducers are required for use.