General Tools DM8215 Dgtl Manomtr 0/15PSI Standard Digital Manometer 30PSI Max Pressure
Representative image only.
General Tools Mfr # DM8215 – Item # WW-97008-77
Ideal for laboratory research equipment, computer peripherals, pneumatic controls, HVAC and more
  • Select from one of 5 pressure units of measure: inH2O, psi, mbar, bar, mmH2O
  • Bright backlight
  • Data hold & record features
  • Min/max button
  • Simple fittings for hose connection
  • Auto power off & low battery indicator
  • 1 year warranty
PRODUCT OPTIONS
₹54,900.00INR / Each
₹27,000.00 - ₹33,700.00INR / Each
for an additional
₹27,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDgtl Manomtr 0/15PSI Standard Digital Manometer 30PSI Max Pressure
  • Warranty1 year
1 Year Warranty
Key Features
  • Select from one of 5 pressure units of measure: inH2O, psi, mbar, bar, mmH2O
  • Bright backlight
  • Data hold & record features
  • Min/max button
  • Simple fittings for hose connection
  • Auto power off & low battery indicator
  • 1 year warranty
More About this Item
Displays gauge or differential pressure.