General Tools DLAF8000 4 in 1 Environmental Airflow Meter
Representative image only.
General Tools Mfr # DLAF8000 – Item # WW-37954-21
Measures airflow, humidity, temperature, and light
  • Displays FPM, MPH, KPH, MPS, knots and temperature
  • One-handed airflow measurement
  • Single and multi-point averages
  • Low-friction ball vane wheels improve accuracy and high and low velocities
  • Min/Max memory, data hold, and F/C switch
  • 1 year warranty
₹44,100.00INR / Each
₹38,900.00 - ₹46,800.00INR / Each
for an additional
₹38,900.00INR / Each
for an additional
₹46,800.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max Air Velocity (ft/min)5910
  • Min Air Velocity (ft/min)80
  • Types Of Probes AcceptedType K thermocouple
  • Min Temperature (° F)32
  • Max Temperature (° F)122
  • Min Temperature (° C)0
  • Max Temperature (° C)50
  • Temperature Accuracy±2.5°F (±1.5°C)
  • Max RH (%)90
  • Min RH (%)10
  • RH Accuracy±4%
  • Description4 in 1 Environmental Airflow Meter
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Displays FPM, MPH, KPH, MPS, knots and temperature
  • One-handed airflow measurement
  • Single and multi-point averages
  • Low-friction ball vane wheels improve accuracy and high and low velocities
  • Min/Max memory, data hold, and F/C switch