Veolia 1236577 Depth CartridFilter, 1.0 micron, plain end, 10"
Representative image only.
Mfr # RO.ZS 01-10/1236577 – Item # WW-29831-63
100% greater dirt-holding capacity lowers total cost of filter operations
  • Up to twice the life of conventional depth filters
  • Up to 50% lower pressure drop
  • 1 year warranty
PRODUCT OPTIONS
₹1,500.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene
  • Micron Rating1
  • Flow Rate3.2 GPM at 1.0 psid
  • Filter Length (in)10
  • Max Temperature (° F)160
  • Max Temperature (° C)71
  • DescriptionGE Depth Cartridge Filter, 1.0 micron, plain end, 10\"
  • Warranty1 year
1 Year Warranty
Key Features
  • Up to twice the life of conventional depth filters
  • Up to 50% lower pressure drop
  • 1 year warranty
More About this Item
Z.Plex® filters are made from thermally bonded fibers of polypropylene. GE Water & Process Technologies certifies that it uses no resin binders, lubricants, antistatic or release agents, or other additives in the manufacture of these cartridges. Suitable to 160°F (71°C).
ROSave.Z® filters
For use in reverse osmosis pretreatment and other pure water applications. Available in 1 and 5 micron pore sizes. The resin used for manufacturing the filter media meets the food contact requirement of the U.S., and is FDA compliant.
Applications
• Bottled water
• Beverage
• Electronics
WellPro.Z® filters
For use in well injection, produced water disposal and other oil and gas applications. Available in 1, 5, 10 and 20 micron pore sizes.
Applications
• Well injection
• Produced water filtration
• Water flood / enhanced oil recovery
• Brine filtration
• Sea water filtration