Freezer, Upright, Basic-use 20.6ft3
Representative image only.
Mfr # FFFU21M1QW – Item # WW-44199-13
Economical models ideal for noncritical storage and general-purpose use
  • Perfect for general-purpose use from the lab to the breakroom
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Freezer Capacity (cu ft)20.5
  • Freezer Capacity (Liters)581
  • Max Temperature - Freezer (° C)-50
  • Power (VAC)220
  • Power (Hz)50
  • Power (Breaker)15 A
  • DescriptionFreezer, Upright, Basic-use 20.6ft3
  • Warranty1 year
1 Year Warranty
Key Features
  • Perfect for general-purpose use from the lab to the breakroom
  • 1 year warranty
More About this Item
These high-quality units provide an economical alternative to lab refrigerators and freezers.
Upright Freezers feature adjustable temperature controls and shelves, interior light, power-on light, and lock with pop-out key for security.