Fowler 54-762-002 Digital Shore Durometer Model
Representative image only.
Fowler Mfr # 54-762-002 – Item # WW-59792-07
Portable Durometers conform to Shore scale and are just right for testing the hardness of flexible materials such as rubber and plastics
  • Manufactured in accordance with DIN, ASTM,ISO and JIS specifications, two styles are available in both analog and digital readout.
  • Applications: 35° blunt taper Durometers are suitable for testing soft and natural rubber products, neoprene, polyesters, soft P.V.C., leather, Thiokol®, nitril rubbers, wax, felt and more. Sharp 30° point Durometers are ideal for testing hard rubber, hard synthetic materials, e.g. acrylics, glass, polystyrol, thermo-plastics, printing rolls, vinyl sheets, cellulose acetates, densified wood, etc.
  • Durometer Bench Stand 53-76-2010 intended for use with 1 kg loading for Shore “A” scales and 5 kg loading for Shore “D” scales. This concept of a low physical force lifting a fixed mass is ideal for both laboratory and production applications. Maximum height of piece to be tested 5.12"/130 mm
SPECS & DESCRIPTION
Specifications & Description
  • TypeD
  • DescriptionSharp 30° point penetrator
Key Features
  • Manufactured in accordance with DIN, ASTM,ISO and JIS specifications, two styles are available in both analog and digital readout.
  • Applications: 35° blunt taper Durometers are suitable for testing soft and natural rubber products, neoprene, polyesters, soft P.V.C., leather, Thiokol®, nitril rubbers, wax, felt and more. Sharp 30° point Durometers are ideal for testing hard rubber, hard synthetic materials, e.g. acrylics, glass, polystyrol, thermo-plastics, printing rolls, vinyl sheets, cellulose acetates, densified wood, etc.
  • Durometer Bench Stand 53-76-2010 intended for use with 1 kg loading for Shore “A” scales and 5 kg loading for Shore “D” scales. This concept of a low physical force lifting a fixed mass is ideal for both laboratory and production applications. Maximum height of piece to be tested 5.12"/130 mm