This item is now discontinued. We recommend the following as suggested replacements: WW-09923-41
Fowler 52-095-007 3-Piece Toolmakers Analog Measuring Set
Representative image only.
Fowler Mfr # 52-095-007 – Item # WW-09923-29
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min range (in)0
  • Max range (in)1
  • ResolutionMicrometer: 0.0001 in; Caliper: 0.001 in
  • AccuracyMicrometer: ±0.0001 in; Caliper: ±0.001 in
  • Description3-Piece Toolmakers Analog Measuring Set
  • Warranty5 Years
5 Year Warranty
Key Features
  • 5 year warranty
More About this Item
Analog Measuring Kit includes micrometer, caliper, flexible 6" rule, and carrying case. Micrometer reads 0 to 1"; 0.0001" graduations; accuracy is ±0.0001". Features friction thimble, locking clamp, and carbide-tipped measuring faces. Caliper measures outside, inside, and depth to 0.100"; 0.001" divisions; accuracy is ±0.001". Features thumbwheel and locking screw.