Fluke 1748/15/EUS Power Quality Logger with 24"/60 cm 1,500A Iflex, Eus Version

Representative image only.
Fluke Mfr # FLUKE-1748/15/EUS – Item # WW-20003-08
Quickly and safely quantify energy usage and analyze power distribution systems
  • Easy access to power quality and energy data in real time
  • Wireless transfer of data minimizes your time in potentially hazardous environments
  • Quickly create standardized reports for analysis
  • Meets IEC 6100-4-30 Class A Edition 3 standard
  • Measures basic power quality parameters, harmonics, interharmonics, transients and waveform deviation events!
  • Includes four iFlex 1500 A probes
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Max AC Voltage (Voltage)1000
  • Max DC Voltage (Voltage)1000
  • Max Frequency Range (KHz)3.5
  • Min Frequency Range (Hz)42.5
  • Total Harmonic Distortion (Thd)10
  • DataloggingInternal flash memory (not user replaceable)
  • MemoryTypical 20 logging sessions of 4 weeks with 1-minute intervals and 500 events
  • Communication PortsEthernet, USB (mini B), and Wi-Fi
  • Battery Type1 3.6V Lithium and 1 1/2 AA (included)
  • Length (in)9 1/8
  • Height (in)2 1/8
  • Width (in)7 1/8
  • Length (cm)23
  • Height (cm)5.4
  • Width (cm)18
  • DescriptionThree-Phase Power Quality Logger, 24\"L iFlex 1500 A probe, Waveform
Two year warrantyConformite Europeenne

Includes

Four 24"L iFlex 1500 A probes, one set (three red and one black) of magnetic probe, four black dolphin clips, set of wire clips, 3-phase + N voltage test lead, set of two 7" (18 cm) L test leads, set of two 6.6 ft (1.5 m) L test leads, USB cable with mini connector, USB drive with software, two Wi-Fi/BLE-to-USB adapters, cable marker kit, soft storage case, magnetic hanger kit, voltage input adapter, and AC power cable.

Key Features

  • Easy access to power quality and energy data in real time
  • Wireless transfer of data minimizes your time in potentially hazardous environments
  • Quickly create standardized reports for analysis
  • Meets IEC 6100-4-30 Class A Edition 3 standard
  • Measures basic power quality parameters, harmonics, interharmonics, transients and waveform deviation events!
  • Includes four iFlex 1500 A probes
  • 2 year warranty

More About this Item

Measure all three voltage and current phases plus the neutral current as well as power and frequency with these power quality loggers. Loggers simultaneously measure up to 500 parameters while capturing events to identify intermittent and hard-to-find power quality issues. Power and power quality variables are automatically logged so you don't have to worry about losing data. Order Fluke model 1746 or 1748 to measure key power quality parameters such as harmonics and interharmonics. Advanced Fluke model 1748 also measures waveform deviations.

Download your data in multiple ways. With both Wi-Fi and Ethernet connection capability, you can use it in hard-to-reach areas as well as in areas where Wi-Fi may be spotty. Loggers feature two external USB ports—one for PC connection and another for quick data download to a USB device. Included software lets you download and quickly analyze measured values and perform advanced analysis to help make decisions in real time. Easily create standardized reports according to commonly used standards such as EN 50160 and IEEE 519.

Easily place in tight spaces and panels—loggers only measure 9-1/8" x 7-1/8" x 2-1/8". Rugged loggers retain IP65 rating when used with IP65 voltage input adapter (included). Safely secure loggers with a standard chain or other security device.

Fluke model 1748 three-phase power quality logger measures voltage, current, power, power factor, frequency, energy forward/reverse, peak demand, THD, flicker, harmonics, interharmonics, unbalance, rapid voltage change events, dips, swells, interruption and transient events tables, mains signaling, inrush current and transients (low frequency)/wave form deviation events.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat