Flir Remote Monitoring Gateway with Four Vibration and Temperature Sensors
Representative image only.
Flir Mfr # SV87-KIT – Item # WW-23029-07
A wireless, connected sensing-solution designed for continuous vibration and temperature monitoring
  • Monitor vibration and temperature to make better-informed decisions
  • Minimize exposure to dangerous environments and difficult-to-access locations
  • Gain quick understanding of the state of manufacturing equipment
  • Check conditions remotely from a mobile device or PC
  • Receive an automatic alert or email the instant a defined vibration or temperature threshold is exceeded
  • Export sensor data to CSV files for analysis
  • Vibration monitoring solution kit
  • 3 year warranty
PRODUCT OPTIONS
₹256,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Vibration Range - Acceleration (g)32
  • Frequency Range10 Hz to 1 kHz
  • Min Temperature (F)-22
  • Max Temperature (F)176
  • Min Temperature (C)-30
  • Max Temperature (C)80
  • Memory32 MB (approx. 5 days of data backup for 4 sensors in case of Wi-Fi outage)
  • DisplayNone
  • DescriptionRemote Monitoring Gateway with Four Vibration and Temperature Sensors
  • Warranty3 Years
3 Year WarrantyConformite Europeenne
Includes
GW65 Gateway, Four Gateway AC connectors for US/EU/UK/AUS power outlets, Four SV87 Remote Vibration/Temperature sensors with adhesive tape, Quick Start Guide, access to configuration and visualization software.
Key Features
  • Monitor vibration and temperature to make better-informed decisions
  • Minimize exposure to dangerous environments and difficult-to-access locations
  • Gain quick understanding of the state of manufacturing equipment
  • Check conditions remotely from a mobile device or PC
  • Receive an automatic alert or email the instant a defined vibration or temperature threshold is exceeded
  • Export sensor data to CSV files for analysis
  • Vibration monitoring solution kit
  • 3 year warranty
More About this Item
The FLIR Vibration Monitoring Solution allows you to monitor machinery for excessive vibration and over-temperature conditions using a mobile device exclusively (stand-alone) or a combination of Windows® PC and mobile device. Accelerometers in the remote sensors measure changes in vibration while thermometers measure surface temperature. This product can check for balance issues, eccentricity, misalignment, and looseness. Vibration trending and analysis can help detect serious problems with industrial machines long before the damage can be seen. Ideal for manufacturing environments that use rotating machinery, industrial pumps, fans, gearboxes, and motors. The remote monitoring gateway stores sensor data and wirelessly transmits readings in real time to a mobile device or PC. This allows professionals to make informed decisions about machine operation and maintenance without a manual inspection.