Flir DM284 Imaging True RMS Industrial Multimeter with IGM

Representative image only.
Flir Mfr # DM284 – Item # WW-20070-00
Locate the exact spot of an electrical problem with the thermal imager
  • Infrared Guided Measurement (IGM™) infrared technology visually guides you to the precise location of a potential electrical problem
  • Work safely—scan a panel or cabinet for hazards using IGM without direct contact
  • Solve problems easier with 18 measurement functions including True RMS, VFD, LoZ, and NCV
  • View thermal and electrical thermocouple measurements simultaneously
  • Select from three color palettes: iron, rainbow, or grayscale
  • 10 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • DC Min (Volts)0
  • DC Max (Volts)1000
  • DC Voltage Accuracy±0.09%
  • DC Max (amps)10
  • AC Min (Volts)0
  • AC Max (Volts)1000
  • Ac Voltage Accuracy±1.0%
  • AC Max (amps)10
  • AC Current Accuracy±1.5%
  • Resistance Max (MΩ)50
  • Capacitance Max (mF)10
  • Capacitance Accuracy1.90%
  • Frequency Max (KHz)99.99
  • Frequency Accuracy0.001
  • ContinuityYes
  • Diode TesterYes
  • True RmsYes
  • Min Temperature (° F)-40
  • Max Temperature (° F)752
  • Min Temperature (° C)-40
  • Max Temperature (° C)400
  • Temperature AccuracyDMM: ±1.0% + 5.4°F (±1.0% + 3°C); IGM: ±1.0% + 9°F (±1.0% + 5°C)
  • Count6000
  • Display2.8\" digital TFT display with bar graph
  • MeasurementMin/Max/Avg
  • Safety RatingCAT III-1000 V, CAT IV-600 V
  • PowerFour AAA batteries or optional rechargeable lithium polymer battery kit
  • Width (in)3.75
  • Length (in)7.875
  • Height (in)2
  • Width (cm)9.5
  • Length (cm)20
  • Height (cm)4.9
  • DescriptionImaging True RMS Industrial Multimeter with IGM
  • Warranty10 Years
10 Year WarrantyIP54 Rated

Includes

Test leads, type K temperature probe, carrying case, AC power adapter/charger, and rechargeable Li-ion battery.

Key Features

  • Infrared Guided Measurement (IGM™) infrared technology visually guides you to the precise location of a potential electrical problem
  • Work safely—scan a panel or cabinet for hazards using IGM without direct contact
  • Solve problems easier with 18 measurement functions including True RMS, VFD, LoZ, and NCV
  • View thermal and electrical thermocouple measurements simultaneously
  • Select from three color palettes: iron, rainbow, or grayscale
  • 10 year warranty

More About this Item

Use this True RMS digital multimeter with thermal imager to quickly troubleshoot any problem areas. The built-in 160 x 120 (19,200 pixels) thermal imager guides you to the precise location of an electrical problem, helping you pinpoint hot spots faster and more efficiently. Use the IGM to scan panels, connectors, and wires without any direct contact, keeping you at a safe distance from dangerous areas. Thermal imager has a temperature range of 14 to 302°F (–10 to 150°C) with 3°C or 3.5% accuracy (whichever is greater). Built-in laser with Class I safety rating has 30:1 distance-to-target ratio with 4 preset emissivity with custom adjustment (0.95 max).

Select from 18 measurement functions to measure and verify any potential electrical issues. Meter also accepts type K thermocouple input and simultaneously displays thermal and electrical thermocouple measurements. Meter features Min, Max, avg, Peak Min, Peak Max, data Hold, and auto power off functions. Built-in flashlight and laser pointer with crosshair targeting help you view and pinpoint difficult-to-measure locations. A convenient tripod mount lets you set up meter on a tripod for long-term measurement. Built-in tilt stand keeps meter upright for easy viewing.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat