Fisher Research Labs FP-ID2100 Magnetic Locator/Metal Detector
Representative image only.
Fisher Research Labs Mfr # FP-ID2100 – Item # WW-99053-02
Affordable, tough, and simple to use while providing maximum performance.
  • Unbeatable performance locating targets near a fence line
  • Pinpoints to 1 inch of center for Valves and Curb boxes
  • Excellent for finding buried ferrous objects including PK nails, iron pipes, valve covers, masonry nails and old hidden culverts
  • Made of strong ABS. Unique end design also protects the switches and the operator is still able to use the controls when wearing thick gloves
  • Pulsating tone/low battery indication and quick, easy battery replacement
₹231,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMagnetic Locator/Metal Detector
Key Features
  • Unbeatable performance locating targets near a fence line
  • Pinpoints to 1 inch of center for Valves and Curb boxes
  • Excellent for finding buried ferrous objects including PK nails, iron pipes, valve covers, masonry nails and old hidden culverts
  • Made of strong ABS. Unique end design also protects the switches and the operator is still able to use the controls when wearing thick gloves
  • Pulsating tone/low battery indication and quick, easy battery replacement
More About this Item
The unique Target Identification System (TID)™ is designed to save time from digging unnecessary holes, especially in hard or frozen ground.
Great for Utilities, Water Work Municipalities, and Construction Road departments to locate manhole covers, catch basins, cast iron pipes, septic tank handles, oil tanks, steel drums, well casings and more. The 2100 can effectively locate valves and curbs up to 9 ft deep, survey markers up to 8 ft deep, and PK nails up to 14'' deep. Can distinguish between a manhole cover and a close iron valve.