Fischer Technology 604-169 Axial single tip probe, Model FD13, 0 to 80 mils
Representative image only.
Fischer Technology Mfr # 604-508 – Item # WW-54100-11
Fischer offers a wide range of probes for a variety of measurement tasks
  • Determine heat damage by means of conductivity measurement
  • Determine ferrite content of steel without destruction
  • Available for sale in the United States, Canada, and Mexico only
  • 6 month warranty
PRODUCT OPTIONS
₹307,000.00INR / Each
₹27,000.00 - ₹33,700.00INR / Each
for an additional
₹27,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Range0 to 80 mil (0 to 2000 µm)
  • AccuracyMagnetic Induction: ±1.5 µm up to 75 µm, ±2% from 75 to 1000 µm, < 3% above 1 mm; Eddy Current: ±1 µm up to 50 µm, ±2% from 50 to 1000 µm, < 3% above 1 mm
  • Measuring MethodEddy current, Magnetic induction
  • ProbeAxial single tip
  • DescriptionAxial single tip probe, Model FD13, 0 to 80 mils
  • Warranty6 month
6 Month Warranty
Key Features
  • Determine heat damage by means of conductivity measurement
  • Determine ferrite content of steel without destruction
  • Available for sale in the United States, Canada, and Mexico only
  • 6 month warranty
More About this Item
Choose the appropriate probe for your application.