Finish Thompson Drum/barrel tube mixer with air-driven motor
Representative image only.
Finish Thompson Mfr # A101125 & DTMS007 – Item # WW-04316-22
Top-to-bottom movement provides thorough mixing without creating a vortex
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Speed (rpm)3000
  • Max Speed (rpm)3000
  • Mounting Type2\" NPT(M)
  • Motor TypeAir-driven
  • Motor hp3/4
  • DescriptionDrum/barrel tube mixer with air-driven motor
  • Warranty3 Months
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
These efficient tube mixers allow you to thoroughly mix the contents of 55-gallon (200 liter) drums without removing the drum lid. The 40" (1016 mm) long mixing tube fits into standard 2" (51 mm) bungs and generates a powerful top-to-bottom flow.
A shaft-mounted turbine inside the mixing tube creates a steady suction effect at four upper intake ports. Mixers draw approximately 13 to 14 gallons (49 to 53 liters) of liquid (750 cp max) per minute through the 2" (51 mm) OD mixing tube. The forcefully expelled liquid resuspends and dissolves settled particles. Choose either an electric or air-drive motor mixer.
Air-drive motor mixer has a 3/4-hp motor that generates speeds from 300 to 3000 rpm with 50-100 psi air input. Mixer includes flow regulator and muffler. We recommend using an air filter/regulator and lubricator.