Extech TP875 Bead Wire Type K Thermocouple Temperature Probe
Representative image only.
Extech Mfr # TP875 – Item # WW-95001-28
Choose the type for your unique application
  • Type J/K and RTD available
  • Applications: FAMEs, pesticides, pharmaceuticals, phenols
  • 1 year warranty
PRODUCT OPTIONS
₹8,400.00INR / Each
₹27,000.00 - ₹33,700.00INR / Each
for an additional
₹27,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBead Wire Type K Thermocouple Temperature Probe
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Type J/K and RTD available
  • Applications: FAMEs, pesticides, pharmaceuticals, phenols
  • 1 year warranty
More About this Item
Special-application thermocouple probes work with any meter accepting standard thermocouple plug connectors. Choose the Type (J/K) compatible with your meter. Each probe is 39" long.
This Type K thermocouple probe measures from -58 to 1000°F (-50 to 538°C) with a subminiature (mini-ANSI) connector.