This item is now discontinued. We recommend the following as suggested replacements: WW-37809-17

Extech RH30 Hygro-Thermometer with Humidity Alert for Desktop or Wall Mount

Representative image only.
Extech Mfr # RH30 – Item # WW-86531-18
Ideal for alerting users of possible mold hazard conditions
  • Monitors relative humidity, temperature, and dew point
  • Sound and light alarm activates when humidity level exceeds user preset
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Min RH Range (%)1
  • Max RH Range (%)99
  • RH Accuracy±3% RH
  • RH Resolution (%)0.001
  • Min Temperature (° F)14
  • Min Temperature (° C)-10
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Temperature Accuracy±1.8°F
  • Temperature Resolution0.1°
  • Display TypeLCD
  • Mounting TypeWall
  • BatteryAA Battery (2)
  • Height (in)3 45/64
  • Width (in)3 45/64
  • Depth (in)29/32
  • DescriptionHygro-Thermometer with Humidity Alert for Desktop or Wall Mount
  • Warranty1 year
1 Year WarrantyIP 67

Key Features

  • Monitors relative humidity, temperature, and dew point
  • Sound and light alarm activates when humidity level exceeds user preset
  • 1 year warranty

More About this Item

The meter warns you of environmental conditions that are out of a desired range with an audible alarm and bright red LED indicator. Features include a super-large, triple display for relative humidity, temperature, and dew point; min/max with reset function; and switchable °F/°C units.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat