Extech PT400 Programmable Outlet Timer

Representative image only.
Extech Mfr # PT400 – Item # WW-35500-08
Save energy on appliances in the home or office
  • Three power settings for added versatility—On, Off, and Auto
  • Save up to ten on/off programs and sixteen combinations for weekdays, weekends, weeks, or each day
  • Easy to install and operate—plugs directly into an existing NA three-prong outlet (Type B)
  • Digital display conveniently shows time, day, and program function—simultaneously
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Timing Capacity99 hours, 59 minutes, 59 seconds
  • Timing Modes12/24 hour mode with Daylight Savings Time function
  • ClockYes
  • Number of Channels1
  • Number of Circuits1
  • Number of Programs16
  • DisplayDigital
  • CaseNo
  • CertificationsFCC, ETL
  • Power (VAC)125
  • Power (Hz)60 Hz
  • Battery TypeInternal rechargeable
  • Battery IncludedYes
  • Height (in)1
  • Depth (in)6
  • Width (in)6
  • Height (cm)2.54
  • Depth (cm)15.24
  • Width (cm)15.24
  • DescriptionProgrammable Outlet Timer
  • Warranty1 year
1 Year Warranty

Key Features

  • Three power settings for added versatility—On, Off, and Auto
  • Save up to ten on/off programs and sixteen combinations for weekdays, weekends, weeks, or each day
  • Easy to install and operate—plugs directly into an existing NA three-prong outlet (Type B)
  • Digital display conveniently shows time, day, and program function—simultaneously
  • 1 year warranty

More About this Item

•Choose between 12- or 24-hour time formats, with or without Daylight Savings Time
•Internal rechargeable battery saves program settings in case of power outage
•Side-access power socket keeps cords out of the way during use
•Use Random function while away—turns on and off for 10 to 31 minutes between 6 PM to 6 AM

•Sixteen Program combinations for individual days or Mon-Fri, Mon-Sun, Sat-Sun, Mon-Sat, Mon/Weds/Fri,
Tues/Thurs/Sat, Mon/Tues/Weds, Thurs/Fri/Sat, Mon/Weds/Fri/Sun
•Rechargeable battery backup lasts more than 100 hours!
•Easy to install and operate
•Programmable count-down function from 99 hours, 59 minutes, and 59 seconds

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat