Extech PQ3470-2 Three-Phase Power Analyzer Kit, 200 A

Representative image only.
Extech Mfr # PQ3470-2 – Item # WW-20046-31
Provides on-screen harmonics display of 1 to 50th order
  • Full system analysis with up to 35 parameters:
  • - V (phase-to-phase), V (phase-to-ground)
  • - A (phase-to-ground)
  • - kW / kVA / kVAR / PF (phase)
  • - kW / kVA / kVAR / PF (system)
  • - kWH / kVAH / kVARH / PFH (system)
  • - Phase angle
  • Select 200 A, 1200 A traditional current clamps or 3000 A flexible current probes
  • Adjustable current transformer CT (1 to 600) and potential transformer PT (1 to 1000) ratio for high power distribution systems
  • Simultaneous display of harmonics and waveform
  • Displays waveform with peak values
  • Analysis of total harmonic distortion (THD)
  • Graphic phase diagram with three-Phase system parameters
  • Three-phase voltage or current unbalanced ratio (VUR, AUR) and unbalanced factor
  • Capture transient events (including dip, swell, and outage) with programmable threshold (%)
  • Wide sampling rate range (2 seconds to 2 hours)
  • Captured measurements can be imported directly into Excel via the SD memory card
  • Built-in clock and calendar
  • Rugged over molded housing
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Max AC Voltage (Voltage)600
  • AC Voltage Accuracy±(0.5% reading)
  • Max AC Current (amps)3000
  • AC Current Accuracy±(0.5% reading)
  • Power Factor Accuracy±0.04
  • Min Frequency Range (Hz)45
  • Max Frequency Range (Hz)65
  • Frequency Accuracy±0.1 Hz
  • Active Power Range9.999 MW
  • Active Power Accuracy±(1.0% reading)
  • Apparent Power Range9.999 MVA
  • Apparent Power Accuracy±(1.0% reading)
  • Reactive Power Range9.999 MVAR
  • Reactive Power Accuracy±(1.0% reading)
  • Active Energy Range 0.0 kWh to 9.999 MWH
  • Active Energy Accuracy±(2.0% reading)
  • Apparent Energy Range0.0 KVAh to 9.999 MVAH
  • Apparent Energy Accuracy±(2.0% reading)
  • Reactive Energy Range0.0 kVARh to 9.999 MVARH
  • Reactive Energy Accuracy±2% reading
  • Power Factor Range0.00 to 1.00
  • Harmonics1 to 50th
  • Harmonics Accuracy±2% (1 to 20); ±4% (21 to 31)
  • Crest Factor1.00 to 99.99
  • Crest Factor Accuracy±(5%)
  • Peak Current Accuracy±(0.5%) AC
  • Peak Voltage Accuracy±(0.5%) AC
  • Phase Angle-180 to +180.0°
  • Phase Accuracy ±1°
  • Datalogging30,000 readings (2 GB SD memory Card)
  • Battery Type8 AA batteries
  • Height (in)8 3/4
  • Width (in)5
  • Depth (in)2 1/2
  • Height (cm)22.2
  • Width (cm)12.7
  • Depth (cm)6.4
  • DescriptionThree-Phase Power Analyzer Kit, 200 A
  • Warranty1 Year
1 Year Warranty

Key Features

  • Full system analysis with up to 35 parameters:
  • - V (phase-to-phase), V (phase-to-ground)
  • - A (phase-to-ground)
  • - kW / kVA / kVAR / PF (phase)
  • - kW / kVA / kVAR / PF (system)
  • - kWH / kVAH / kVARH / PFH (system)
  • - Phase angle
  • Select 200 A, 1200 A traditional current clamps or 3000 A flexible current probes
  • Adjustable current transformer CT (1 to 600) and potential transformer PT (1 to 1000) ratio for high power distribution systems
  • Simultaneous display of harmonics and waveform
  • Displays waveform with peak values
  • Analysis of total harmonic distortion (THD)
  • Graphic phase diagram with three-Phase system parameters
  • Three-phase voltage or current unbalanced ratio (VUR, AUR) and unbalanced factor
  • Capture transient events (including dip, swell, and outage) with programmable threshold (%)
  • Wide sampling rate range (2 seconds to 2 hours)
  • Captured measurements can be imported directly into Excel via the SD memory card
  • Built-in clock and calendar
  • Rugged over molded housing
  • 1 year warranty

More About this Item

This graphical power and harmonics analyzer allows you to easily capture data for analyzing power systems. The analyzer logs up to 30,000 readings on a removable SD memory card in Microsoft® Excel® format and features an intuitive menu-driven interface that makes set up and operation quick and simple.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat