Extech IR300UV Infrared Thermometer with UV Refrigerant Leak Detector and LED Flashlight
Representative image only.
Extech Mfr # IR300UV – Item # WW-90441-00
Maintain accurate measurement distance with coaxial guidance laser
  • Take quick noncontact temperature readings on unsafe surfaces
  • Easily reveal refrigerant leaks on pipes, AC units, and automotives
  • No need to work in the dark—built-in flashlight provides a bright LED light source
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Distance To Target Size Ratio (D:S)12:1
  • EmissivityAdjustable
  • Sighting TypeCircular laser
  • Laser ClassClass II
  • Min Temperature (° F)-22
  • Max Temperature (° F)932
  • Min Temperature (° C)-30
  • Max Temperature (° C)500
  • Accuracy±1.5% of reading or ±3°F (±1.5°C), whichever is greater
  • Resolution0.2°F (0.1°C)
  • Display TypeBacklit LCD
  • Low Battery IndicatorYes
  • BatteryAA Battery (3)
  • DescriptionInfrared Thermometer with UV Refrigerant Leak Detector and LED Flashlight
  • Warranty3 year
3 Year WarrantyConformite EuropeenneLaser Class II
Key Features
  • Take quick noncontact temperature readings on unsafe surfaces
  • Easily reveal refrigerant leaks on pipes, AC units, and automotives
  • No need to work in the dark—built-in flashlight provides a bright LED light source
  • 3 year warranty
More About this Item
Versatile IR thermometer with laser sighting is ideal for taking quick temperature readings of surfaces that are hard to reach or too hot to get close. Use the UV detector in combination with standard UV dye (not included) to reveal refrigerant leaks; detector features five blue UV LEDs (460 to 470 nm). Convenient built-in flashlight with five white LEDs lets you work in dark areas without requiring another light source.
Adjustable emissivity provides better accuracy when measuring different surfaces. Meter features programmable high and low temperature set points with audible and visual alarm. Functions include Min, Max, Avg, Max–Min, and auto power-off. Simply release the trigger to hold a reading.