Extech EX505-K Heavy Duty Industrial Multimeter Kit
Representative image only.
Extech Mfr # EX505-K – Item # WW-20034-47
Save time and money with these multifunctional kits!
  • Kit Includes:
  • EX505 industrial grade, CAT IV-600 V TRMS, 4000 count display, waterproof, drop-proof, rugged multimeter with True RMS functions
  • DV25 dual range AC voltage detector (24 to 1000 V AC / 100 to 1000 V AC) plus flashlight with audible/ visual indication, double-molded housing, and CAT IV-1000 V safety rating
  • TL808 eight piece professional test lead kit with CAT III-1000 V safety rating
  • Set of double molded professional test leads
  • Type K thermocouple bead probe and Type K to Banana Input Adaptor
  • Supplied in a soft carrying case that provides protection and organization for the meters whenever they are needed
  • Dimensions: 9.8 x 8 x 2'' (248 x 203 x 51 mm)
  • 1 year warranty
₹32,200.00INR / Each
₹20,200.00 - ₹27,000.00INR / Each
for an additional
₹20,200.00INR / Each
for an additional
₹27,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DC Max (Volts)600
  • AC Max (Volts)600
  • Safety RatingCAT IV 600V, EN 61010-1
  • DescriptionHeavy Duty Industrial Multimeter Kit
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • Kit Includes:
  • EX505 industrial grade, CAT IV-600 V TRMS, 4000 count display, waterproof, drop-proof, rugged multimeter with True RMS functions
  • DV25 dual range AC voltage detector (24 to 1000 V AC / 100 to 1000 V AC) plus flashlight with audible/ visual indication, double-molded housing, and CAT IV-1000 V safety rating
  • TL808 eight piece professional test lead kit with CAT III-1000 V safety rating
  • Set of double molded professional test leads
  • Type K thermocouple bead probe and Type K to Banana Input Adaptor
  • Supplied in a soft carrying case that provides protection and organization for the meters whenever they are needed
  • Dimensions: 9.8 x 8 x 2'' (248 x 203 x 51 mm)