Extech ET25 Compact AC/DC Voltage Tester, 80-250V
Representative image only.
Extech Mfr # ET25 – Item # WW-20021-24
Detect voltage quickly and safely
  • Voltage detection from 80 to 250 V AC/DC
  • Neon indicator glows if circuit is live
  • Insulated housing provides added safety
  • Built-in pocket clip keeps unit secure and readily available
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DC Min (Volts)80
  • DC Max (Volts)250
  • AC Min (Volts)80
  • AC Max (Volts)250
  • DescriptionCompact AC/DC Voltage Tester, 90-300V
  • Warranty1 Year
1 Year Warranty
Key Features
  • Voltage detection from 80 to 250 V AC/DC
  • Neon indicator glows if circuit is live
  • Insulated housing provides added safety
  • Built-in pocket clip keeps unit secure and readily available
  • 1 year warranty
More About this Item
Safely detect the presence of voltage quickly and easily with the ET series of electrical testers. Visible and audible alarms notify you if live voltage is nearby and should be avoided or powered down. Choose from a wide selection of compact voltage and continuity testers for use in residential, commercial, and industrial applications.