Extech EN100-TP Optional RTD Temperature Probe (Pt-1000 ohm)
Representative image only.
Extech Mfr # EN100-TP – Item # WW-10000-83
Choose the type for your unique application
  • Type J/K and RTD available
  • Applications: FAMEs, pesticides, pharmaceuticals, phenols
  • 1 year warranty
PRODUCT OPTIONS
₹8,200.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionOptional RTD Temperature Probe (Pt-1000 ohm)
  • Warranty1 year
1 Year Warranty
Key Features
  • Type J/K and RTD available
  • Applications: FAMEs, pesticides, pharmaceuticals, phenols
  • 1 year warranty
More About this Item
Special-application thermocouple probes work with any meter accepting standard thermocouple plug connectors. Choose the Type (J/K) compatible with your meter. Each probe is 39" long.