This item is now discontinued. We recommend the following as suggested replacements: WW-20034-53
Extech DV20 AC Voltage Detector/Flashlight
Representative image only.
Extech Mfr # DV20 – Item # WW-20021-19
Non-contact detection of AC Voltage with built-in flashlight
  • Non-Contact Detection of AC Voltage
  • For use on 50/60Hz circuits
  • Tip fits into outlets or against wire insulation
  • Flashing red LED light and push button ON/OFF
  • Locate breaks in power cables
  • Convenient size with pocket clip
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • AC Min (Volts)100
  • AC Max (Volts)600
  • Powertwo AAA batteries
  • DescriptionAC Voltage Detector/Flashlight
  • Warranty1 Year
1 Year Warranty
Key Features
  • Non-Contact Detection of AC Voltage
  • For use on 50/60Hz circuits
  • Tip fits into outlets or against wire insulation
  • Flashing red LED light and push button ON/OFF
  • Locate breaks in power cables
  • Convenient size with pocket clip