Extech 42515 Infrared Thermometer with Type K Thermocouple Input (13: 1)
Representative image only.
Extech Mfr # 42515 – Item # WW-90415-15
Measures both non-contact and contact temperature
  • Wide temperature range for IR temperature and type K thermocouple measurements
  • Automatic emissivity adjustment (for temperatures 212°F or higher)
  • Memory stores up to 20 readings
  • Adjustable high/low visual and audible alarm
  • MAX/MIN/AVG/DIF features display highest, lowest, average, and MAX minus MIN values
  • Large LCD display with bright backlight
  • Built-in laser pointer improves aim
  • Data Hold, Auto power off, and Low battery indication
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Distance To Target Size Ratio (D:S)13:1
  • EmissivityAdjustable
  • Sighting TypeSingle-point laser
  • Min Temperature (° F)-58
  • Max Temperature (° F)1475
  • Min Temperature (° C)-50
  • Max Temperature (° C)800
  • AccuracyInfrared: ±2% of reading or ±4.0°F (2.0°C), whichever is greater; Contact: ±1.5% reading ± 2.0°F (1.0°C)
  • Resolution0.1°
  • Probe TypeType-K
  • Probe Min Temperature (° F)-58
  • Probe Max Temperature (° F)2498
  • Probe Min Temperature (° C)-50
  • Probe Max Temperature (° C)1370
  • Display Type4000 count backlit LCD
  • Battery9v Battery (1)
  • Temperature InputYes
  • DescriptionInfrared Thermometer with Type K Thermocouple Input (13: 1)
  • Warranty1 year
1 Year Warranty
Key Features
  • Wide temperature range for IR temperature and type K thermocouple measurements
  • Automatic emissivity adjustment (for temperatures 212°F or higher)
  • Memory stores up to 20 readings
  • Adjustable high/low visual and audible alarm
  • MAX/MIN/AVG/DIF features display highest, lowest, average, and MAX minus MIN values
  • Large LCD display with bright backlight
  • Built-in laser pointer improves aim
  • Data Hold, Auto power off, and Low battery indication
  • 1 year warranty
More About this Item
Cole-Parmer provides a wide offering of Professional Infrared Thermometers with varying features and accuracies depending on your application.
We now offer an FDA-cleared infrared thermometer.
Our other infrared thermometers are not approved for medical usage and are not FDA-approved.
For help in selecting an infrared thermometer please contact the Technical Support team at Cole-Parmer.