Excel Scientific AlumnaSeal 96™ Sealing Film, 96-Well Plates, Aluminum Foil, Non-Sterile, 38µm; 100/PK
Representative image only.
Excel Scientific Mfr # F-96-100 – Item # WW-13024-82
Specially formulated for cold storage to -80 °C
  • Certified free of DNase-, RNase-, and nucleic-acids
  • Excellent barrier properties to retard evaporation
  • Easily pierceable with pipet tips or robotic probes for sample recovery
  • 3 year warranty
PRODUCT OPTIONS
₹11,900.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • SterileNo
  • Min Temperature (° C)-80
  • Max Temperature (° C)120
  • MaterialNonpermeable aluminum foil
  • Width (mm)127
  • Length (mm)77.8
  • DescriptionThermal Cycling Film, Aluminum Seal, 96-Well Plates, 38um; 100/Pk
  • Warranty3 year
3 Year Warranty
Key Features
  • Certified free of DNase-, RNase-, and nucleic-acids
  • Excellent barrier properties to retard evaporation
  • Easily pierceable with pipet tips or robotic probes for sample recovery
  • 3 year warranty
More About this Item
AlumaSeal CS sealing films are specially formulated 50 µm aluminum foil sealing films with a 50 µm layer of acrylic adhesive that withstands cold storage at temperatures to –80°C. AlumaSeal CS sealing films are not recommended for PCR or thermocycling. A single non-perforated end tab simplifies application. Simply hold the tab and strip the backing from the body of the sealing film as it lays on the plate to avoid curling.