Excel Scientific RL-PLT-01 Microplate Sealing Film Plate Roller, Soft Rubber, 4"; 1/Bx
Representative image only.
Excel Scientific Mfr # RL-PLT-01 – Item # WW-13024-99
Easily adhere sealing films to microplates
  • Optimize performance of your microplate applications with these accessories
PRODUCT OPTIONS
₹7,300.00INR / Each
Often Bought with…
Excel Scientific AlumaSeal Sealing Film
11,900.00 - ₹15,500.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Plate TypeSealing Film Plate Roller
  • MaterialSoft Rubber
  • Width (in)4
  • DescriptionMicroplate Sealing Film Plate Roller, Soft Rubber, 4\"; 1/Bx
Key Features
  • Optimize performance of your microplate applications with these accessories
More About this Item
Ensure a secured seal of your sealing film with these accessories.
Complete the application of AlumaSeal 384 sealing films by pressing with the accessory plate roller. This assures a secure uniform seal around all wells.
Compatible with sealing film 13024-16 and 13024-18. Sealing film sold separately.