Eurotherm AP4382-2000-1 Bipolar Isolator Dc Voltage/ Current Input Voltage Output; 120/240 VAC, 50-60 Hz
Representative image only.
Eurotherm Mfr # AP4382-2000-1 – Item # WW-68478-02
Field configurable - compact design
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Output ch1 (VDC)10
  • Min Output ch1 (VDC)-10
  • Power Supply (VAC)120/240
  • DescriptionBipolar Isolator Dc Voltage/ Current Input Voltage Output
  • Warranty5 years
5 Year Warranty
Key Features
  • 5 year warranty
More About this Item
Limit Alarms are setpoint controllers that regulate industrial processes, whether discrete, batch or continuous. Alarms are indicators of problems, be they production line faults, process upsets, equipment failures or control system abnormalities.