Eppendorf 940002001 Electroporation Buffer for Eukaryotic Cells, Hypo-osmolar, 100 mL
Representative image only.
Eppendorf Mfr # 940002001 – Item # WW-36205-60
Provides quality transfection results
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)100
  • ColorClear
  • DescriptionElectroporation Buffer for Eukaryotic Cells, Hypoosmolar, 100 mL
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The best possible transfection results are obtained by using the original buffers from Eppendorf®, which have been tested for sterility as well as for the absence of mycoplasma, endotoxins, and pyrogens, and have undergone a thorough cytotoxicity test.
The hypo-osmolar buffer is for electroporation, and increases transfection efficiency, particularly for mammalian cells. It allows the electrode formation of the cells and, as a result, easier membrane penetration. The sodium-free buffer system is adapted to the inner cell environment and stabilizes the Na+/K+ gradient across the cell membrane, so even highly sophisticated transfection experiments are possible.