Environmental Express Vial, Amber Econo CT PTFE Liner, 60mL, 144/C
Representative image only.
Environmental Express Item # WW-53100-17
Borosilicate glass vials meet EPA guidelines for contamination-free containers
  • Choose between 40- and 60-mL capacities
  • Include standard screw-top caps
PRODUCT OPTIONS
₹26,100.00INR / Case of 144
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)60
  • Tube MaterialBorosilicate
  • Tube Bottom ShapeFlat
  • Tube ColorAmber
  • Cap TypePTFE-lined screw-top
  • Cap MaterialPTFE-lined screw-top
  • Cap colorWhite
  • Thread24 mm
  • SterileNo
  • AutoclavableNo
  • Level1
  • DescriptionVial, Amber Econo CT PTFE Liner, 60mL, 144/C
Key Features
  • Choose between 40- and 60-mL capacities
  • Include standard screw-top caps
More About this Item
Level 3 VOA vials are not certified and ideal for general use containers that meet EPA recommended guidlines. Vials come with closed-top caps with PTFE liner. Choose from clear vials to easily view samples or amber vials to protect light-sensitive samples.