Environmental Express Tubing and Fitting Sets, for Cyanide Glassware Set, 10/ST
Representative image only.
Environmental Express Item # WW-35209-68
Replacement components for the MicroBlock systems
  • Reduce chemical waste by 90%
  • Get more accurate distillations
  • Comply with all EPA methodologies for the referenced methods; suitable for cyanide (EPA 335.4), phenol (EPA 420.4) and ammonia (EPA 350.1)
  • 3 month warranty
PRODUCT OPTIONS
₹187,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialBorosilicate Glass
  • Compatible WithMicroBlock V Systems (35209-50, 35209-51, or 35209-54)
  • DescriptionTubing and Fitting Sets, for Cyanide Glassware Set, 10/ST
  • Warranty3 month
3 Month Warranty
Key Features
  • Reduce chemical waste by 90%
  • Get more accurate distillations
  • Comply with all EPA methodologies for the referenced methods; suitable for cyanide (EPA 335.4), phenol (EPA 420.4) and ammonia (EPA 350.1)
  • 3 month warranty
More About this Item
Minimize heat loss and maintain uniform distillations by using specially designed MicroBlock glassware. Cap distillation tubes after distillation to prevent evaporation or contamination.
Tubing & Quick Connect fittings for 10 sets of cyanide distillation glasware.