Environmental Express SE Standard, Indium, 1000ppm, 100mL - HAZMAT
Representative image only.
Environmental Express Item # WW-15962-34
Ensure consistent results-standards are quality checked at four different steps
  • Quality ingredients produce quality standards, eliminating discrepancies in your lab
  • Our single element standards are certified by ICP-MS or ICP-OES to levels below parts-per-billion
  • Certificate of analysis shows trace levels of 68 metals down to picogram levels
  • Manufactured to exact specifications in an environment that is ISO 9001:2008 certified and 17025:2005 accredited as well as ISO Guide 34 certified
  • Special formulations-we can create custom blends in any volume you need, from 50 mL to 100 L
SPECS & DESCRIPTION
Specifications & Description
Key Features
  • Quality ingredients produce quality standards, eliminating discrepancies in your lab
  • Our single element standards are certified by ICP-MS or ICP-OES to levels below parts-per-billion
  • Certificate of analysis shows trace levels of 68 metals down to picogram levels
  • Manufactured to exact specifications in an environment that is ISO 9001:2008 certified and 17025:2005 accredited as well as ISO Guide 34 certified
  • Special formulations-we can create custom blends in any volume you need, from 50 mL to 100 L