Environmental Express SC600 Disposable Digestion Tube, with Hinged Lid, 100 mL; 250/Pack
Representative image only.
Environmental Express Item # WW-53100-05
Disposable digestion tubes eliminate additional glassware washing steps
  • Convenient, disposable tubes
  • Model number SC600
₹20,800.00INR / Pkg of 250
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)100
  • Tube MaterialPolypropylene
  • Tube ColorTranslucent
  • Cap TypeSnap-Cap
  • Cap MaterialPolypropylene
  • DescriptionDisposable Digestion Tube, with Hinged Lid, 100 mL; 250/Pack
Key Features
  • Convenient, disposable tubes
  • Model number SC600
More About this Item
These polypropylene, snap digestion vessels with attached lid eliminate the need for glassware cleanup. All digestion vessels have molded in graduations and serve as graduated cylinders as well as sample storage containers. This vessel does not fit into Environmental Express HotBlock™.