Environmental Express QC-TMFM-X 7300 Trace Metals Filters, 10 to 100 µg, 37 mm; 10 filters, 5 blanks
Representative image only.
Environmental Express Item # WW-35209-02
Added quality control to your industrial hygiene testing
  • Meets NIOSH Method 7300 requirements
  • Industrial hygiene standards for trace metals
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Description7300 Trace Metals Filters, 10 to 100 µg, 37 mm; 10 filters, 5 blanks
  • Warranty3 month
3 Month Warranty
Key Features
  • Meets NIOSH Method 7300 requirements
  • Industrial hygiene standards for trace metals
  • 3 month warranty
More About this Item
Filters are 37-mm diameter mixed cellulose ester (MCE) with a 0.8 µm pore size. The concentrations are reported as µg per filter, and are certified to ±1%. Expiration date of one year.
Trace metal in µg per filter
Aluminum -
Arsenic 100
Barium 25
Beryllium 25
Cadmium 25
Chromium 25
Cobalt 25
Copper 50
Iron 50
Lead 50
Maganese 25
Nickel 25
Silver 10
Thalium 25
Vanadium 25
Zinc 100
Uranium -
Available for sale in the United States, UK, and India only