Environmental Express FilterMate™ 50 mL Digestion Tube Holder
Representative image only.
Environmental Express Item # WW-35202-06
No need for a brand new FilterMate
  • Plunger or sample tube available
  • Works with 50 mL sample tube and FilterMate
  • 3 month warranty
PRODUCT OPTIONS
₹14,500.00INR / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHDPE
  • Compatible With50 mL digestion tubes
  • Description50 mL Digestion Tube Holder
  • Warranty3 month
3 Month Warranty
Key Features
  • Plunger or sample tube available
  • Works with 50 mL sample tube and FilterMate
  • 3 month warranty
More About this Item
The machined HDPE holder adds stability when performing FilterMate filtrations. The 50 mL sample tube is placed into the center hole of the FilterMate holder prior to filtration. The holder secures the digestion tube as the FilterMate is depressed through the liquid sample.
Holder for FilterMater filtrations, secures a 50 mL digestion tube during filtration. This machined HDPE fixture adds stability when performing FilterMate filtrations. The sample tube is placed into the center hole of the FilterMate holder prior to filtration. The holder secures the digestion tube as the FilterMate is depressed through the liquid sample.