Environmental Express Filter Membrane, Polypropylene, 0.1 um, 25 mm dia., 100/PK
Representative image only.
Environmental Express Item # WW-35200-13
Prevent sample-to-sample contamination
  • Resistant to strong acids and bases
PRODUCT OPTIONS
₹16,300.00INR / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Pore Size (µm)0.1
  • Diameter (mm)25
  • DescriptionFilter Membrane, Polypropylene, 0.1 um, 25 mm dia
Key Features
  • Resistant to strong acids and bases
More About this Item
Ideal replacement membrane filters for your radiological extration filter funnels and SimpleDist micro tubes.
Polypropylene filters for alpha particle collection are resistant to strong acids and bases. Use in the co-precipitation technique. Each lot of material is qualified by Environmental Express to ensure proper collection efficiency.